missing translation for 'onlineSavingsMsg'
Learn More

1-(2,6-Difluorophenyl)ethanol, 97%, Thermo Scientific Chemicals

Catalog No. p-7031629
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3277109 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH3277109 Supplier Thermo Scientific Chemicals Supplier No. H3277109

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 87327-65-9 | Molecular Formula: C8H8F2O | Molecular Weight: 158.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 87327-65-9
Molecular Formula C8H8F2O
Molecular Weight (g/mol) 158.15
MDL Number MFCD00274321
InChI Key SIYWDKQSSDBLOA-UHFFFAOYNA-N
Synonym 1-2,6-difluorophenyl ethanol, 1-2,6-difluorophenyl ethan-1-ol, 2,6-difluoro-alpha-methylbenzyl alcohol, benzenemethanol,2,6-difluoro-a-methyl, 2,6-difluoro-.alpha.-methylbenzyl alcohol
PubChem CID 5012075
IUPAC Name 1-(2,6-difluorophenyl)ethanol
SMILES CC(O)C1=C(F)C=CC=C1F

Specifications

Refractive Index 1.486
Quantity 10 g
Formula Weight 158.15
Percent Purity 97%
Chemical Name or Material 1-(2,6-Difluorophenyl)ethanol
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only