missing translation for 'onlineSavingsMsg'
Learn More

1,2,4-Trihydroxybenzene 98.0+%, TCI America™

Catalog No. H024910G
Change view
Click to view available options
Quantity:
10 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H024910G 10 g
H024925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H024910G Supplier TCI America Supplier No. H024910G

Chemical Identifiers

CAS 533-73-3
Molecular Formula C6H6O3
Molecular Weight (g/mol) 126.111
MDL Number MFCD00002198
InChI Key GGNQRNBDZQJCCN-UHFFFAOYSA-N
Synonym 1,2,4-benzenetriol, 1,2,4-trihydroxybenzene, hydroxyhydroquinone, hydroxyquinol, oxyhydroquinone, 2,5-dihydroxyphenol, oxyhydrochinon, hydroquinone, hydroxy, 4-hydroxycatechol, 1,3,4-trihydroxybenzene
PubChem CID 10787
ChEBI CHEBI:16971
IUPAC Name benzene-1,2,4-triol
SMILES C1=CC(=C(C=C1O)O)O

Specifications

Melting Point 140°C
Quantity 10 g
Formula Weight 126.11
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,2,4-Trihydroxybenzene
RTECSNumber DC4200000
TSCA Yes