accessibility menu, dialog, popup

UserName

CAS RN 533-73-3

IUPAC Name: benzene-1,2,4-triol
Synonyms: 1,2,4-benzenetriol hydroxyquinol oxyhydrochinon
Molecular Weight (g/mol): 126.11
Molecular Formula: C6H6O3
InChi Key: GGNQRNBDZQJCCN-UHFFFAOYSA-N
SMILES: OC1=CC=C(O)C(O)=C1
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)