missing translation for 'onlineSavingsMsg'
Learn More

1,2,4-Trihydroxybenzene, 97%

Catalog No. AAA1306903
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1306903 1 g
AAA1306906 5 g
AAA1306914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1306903 Supplier Thermo Scientific Chemicals Supplier No. A1306903
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 533-73-3 | C6H6O3 | 126.111 g/mol

1,2,4-Trihydroxybenzene is an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.It is used as a metabolite of benzene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,2,4-Trihydroxybenzene is an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.It is used as a metabolite of benzene.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from acid chlorides, acid anhydrides, bases, oxidizing agents. Air & Light Sensitive.

Chemical Identifiers

CAS 533-73-3
Molecular Formula C6H6O3
Molecular Weight (g/mol) 126.111
MDL Number MFCD00002198
InChI Key GGNQRNBDZQJCCN-UHFFFAOYSA-N
Synonym 1,2,4-benzenetriol, 1,2,4-trihydroxybenzene, hydroxyhydroquinone, hydroxyquinol, oxyhydroquinone, 2,5-dihydroxyphenol, oxyhydrochinon, hydroquinone, hydroxy, 4-hydroxycatechol, 1,3,4-trihydroxybenzene
PubChem CID 10787
ChEBI CHEBI:16971
IUPAC Name benzene-1,2,4-triol
SMILES C1=CC(=C(C=C1O)O)O

Specifications

Melting Point ∼140°C (sublimation)
Quantity 1 g
Beilstein 2042863
Sensitivity Air and light sensitive
Merck Index 14,1073
Solubility Information Soluble in water.
Formula Weight 126.11
Percent Purity 97%
Chemical Name or Material 1,2,4-Trihydroxybenzene
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 208-575-1
RTECSNumber DC4200000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only