missing translation for 'onlineSavingsMsg'
Learn More

1,2,3-Trifluorobenzene 98.0+%, TCI America™

Catalog No. p-7137176
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T23561G 1 g
T23565G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T23561G Supplier TCI America Supplier No. T23561G
Only null left

Chemical Identifiers

CAS 1489-53-8
Molecular Formula C6H3F3
Molecular Weight (g/mol) 132.085
MDL Number MFCD00012013
InChI Key AJKNNUJQFALRIK-UHFFFAOYSA-N
Synonym trifluorobenzene, benzene, trifluoro, benzene, 1,2,3-trifluoro, 3,4,5-trifluoro-benzene, pubchem1067, 1,2,3-fluorobenzene, 1,2,3-trifluorobenze, 1,2,3-trifluorbenzen, 1,2,6-trifluorobenzene, 2,3,4-trifluorobenzene
PubChem CID 15147
IUPAC Name 1,2,3-trifluorobenzene
SMILES C1=CC(=C(C(=C1)F)F)F

Specifications

Color Yellow
Boiling Point 95°C
Quantity 1 g
UN Number 1993
Formula Weight 132.09
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,2,3-Trifluorobenzene