accessibility menu, dialog, popup

UserName

FFF

CAS RN 1489-53-8

IUPAC Name: 1,2,3-trifluorobenzene
Synonyms: 1,2,3-trifluorobenzene
Molecular Weight (g/mol): 132.09
Molecular Formula: C6H3F3
InChi Key: AJKNNUJQFALRIK-UHFFFAOYSA-N
SMILES: FC1=CC=CC(F)=C1F
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)