missing translation for 'onlineSavingsMsg'
Learn More

1,2,3-Trifluorobenzene, 98+%

Catalog No. AAB2205503
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2205503 1 g
AAB2205506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2205503 Supplier Thermo Scientific Chemicals Supplier No. B2205503
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1489-53-8 | C6H3F3 | 132.085 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1489-53-8
Molecular Formula C6H3F3
Molecular Weight (g/mol) 132.085
MDL Number MFCD00012013
InChI Key AJKNNUJQFALRIK-UHFFFAOYSA-N
Synonym trifluorobenzene, benzene, trifluoro, benzene, 1,2,3-trifluoro, 3,4,5-trifluoro-benzene, pubchem1067, 1,2,3-fluorobenzene, 1,2,3-trifluorobenze, 1,2,3-trifluorbenzen, 1,2,6-trifluorobenzene, 2,3,4-trifluorobenzene
PubChem CID 15147
IUPAC Name 1,2,3-trifluorobenzene
SMILES C1=CC(=C(C(=C1)F)F)F

Specifications

Density 1.28
Boiling Point 94°C to 95°C
Flash Point −3°C (26°F)
Refractive Index 1.422
Quantity 1 g
UN Number UN2387
Beilstein 1862388
Formula Weight 132.09
Percent Purity ≥98%
Chemical Name or Material 1,2,3-Trifluorobenzene
Hazard Category H225-H315-H319-H335
Hazard Statement GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLUOROBENZENE
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only