missing translation for 'onlineSavingsMsg'
Learn More

1,2,3-Tribromobenzene 98.0+%, TCI America™

Catalog No. p-7137701
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T33291G 1 g
T33295G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T33291G Supplier TCI America Supplier No. T33291G
Only null left

Chemical Identifiers

CAS 608-21-9
Molecular Formula C6H3Br3
Molecular Weight (g/mol) 314.80
MDL Number MFCD00192668
InChI Key GMVJKSNPLYBFSO-UHFFFAOYSA-N
PubChem CID 11842
IUPAC Name 1,2,3-tribromobenzene
SMILES BrC1=CC=CC(Br)=C1Br

Specifications

Melting Point 88°C
Boiling Point 283°C
Quantity 1 g
Formula Weight 314.80
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,2,3-Tribromobenzene