accessibility menu, dialog, popup

UserName

BrBrBr

CAS RN 608-21-9

IUPAC Name: 1,2,3-tribromobenzene
Synonyms: 1,2,3-tribromobenzene
Molecular Weight (g/mol): 314.8
Molecular Formula: C6H3Br3
InChi Key: GMVJKSNPLYBFSO-UHFFFAOYSA-N
SMILES: BrC1=CC=CC(Br)=C1Br
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results