missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,2,3,6-Tetrahydro-4-phenylpyridine Hydrochloride 98.0+%, TCI America™
Supplier: TCI America T135625G
Specifications
| 1,2,3,6-Tetrahydro-4-phenylpyridine Hydrochloride | |
| 203°C | |
| C11H14ClN | |
| 25 g | |
| 4-phenyl-1,2,3,6-tetrahydropyridine hydrochloride, 1,2,3,6-tetrahydro-4-phenylpyridine hydrochloride, 4-phenyl-1,2,3,6-tetrahydroxpyridine, pyridine, 1,2,3,6-tetrahydro-4-phenyl-, hydrochloride, 4-phenyl-1,2,5,6-tetrahydropyridine, chloride, acmc-209jsh, 4-phenyl-1,2,3,6-tetra-hydropyridine, 4-phenyl-1,2,3,6-tetrahydropyridine hcl, 4-phenyl-1,2,3,6-tetrahydropyridinehydrochloride, 4-phenyl-1,2,3,6-tetrahydro-pyridine hydrochloride | |
| C1CNCC=C1C2=CC=CC=C2.Cl | |
| 195.69 | |
| 195.69 | |
| Crystalline Powder |
| 43064-12-6 | |
| White-Yellow | |
| MFCD00012752 | |
| 2811 | |
| POGWXTJNUCZEPR-UHFFFAOYSA-N | |
| 4-phenyl-1,2,3,6-tetrahydropyridine;hydrochloride | |
| 2723860 | |
| ≥98.0% (HPLC,T) |
Chemical Identifiers
| 43064-12-6 | |
| 195.69 | |
| POGWXTJNUCZEPR-UHFFFAOYSA-N | |
| 2723860 | |
| C1CNCC=C1C2=CC=CC=C2.Cl |
| C11H14ClN | |
| MFCD00012752 | |
| 4-phenyl-1,2,3,6-tetrahydropyridine hydrochloride, 1,2,3,6-tetrahydro-4-phenylpyridine hydrochloride, 4-phenyl-1,2,3,6-tetrahydroxpyridine, pyridine, 1,2,3,6-tetrahydro-4-phenyl-, hydrochloride, 4-phenyl-1,2,5,6-tetrahydropyridine, chloride, acmc-209jsh, 4-phenyl-1,2,3,6-tetra-hydropyridine, 4-phenyl-1,2,3,6-tetrahydropyridine hcl, 4-phenyl-1,2,3,6-tetrahydropyridinehydrochloride, 4-phenyl-1,2,3,6-tetrahydro-pyridine hydrochloride | |
| 4-phenyl-1,2,3,6-tetrahydropyridine;hydrochloride |
Safety and Handling
TSCA : No