missing translation for 'onlineSavingsMsg'
Learn More

1,2,3,6-Tetrahydro-4-phenylpyridine Hydrochloride 98.0+%, TCI America™

Catalog No. p-7136731
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T13565G 5 g
T135625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T13565G Supplier TCI America Supplier No. T13565G

Chemical Identifiers

CAS 43064-12-6
Molecular Formula C11H14ClN
Molecular Weight (g/mol) 195.69
MDL Number MFCD00012752
InChI Key POGWXTJNUCZEPR-UHFFFAOYSA-N
Synonym 4-phenyl-1,2,3,6-tetrahydropyridine hydrochloride, 1,2,3,6-tetrahydro-4-phenylpyridine hydrochloride, 4-phenyl-1,2,3,6-tetrahydroxpyridine, pyridine, 1,2,3,6-tetrahydro-4-phenyl-, hydrochloride, 4-phenyl-1,2,5,6-tetrahydropyridine, chloride, acmc-209jsh, 4-phenyl-1,2,3,6-tetra-hydropyridine, 4-phenyl-1,2,3,6-tetrahydropyridine hcl, 4-phenyl-1,2,3,6-tetrahydropyridinehydrochloride, 4-phenyl-1,2,3,6-tetrahydro-pyridine hydrochloride
PubChem CID 2723860
IUPAC Name 4-phenyl-1,2,3,6-tetrahydropyridine;hydrochloride
SMILES C1CNCC=C1C2=CC=CC=C2.Cl

Specifications

Melting Point 203°C
Color White-Yellow
Quantity 5 g
UN Number 2811
Formula Weight 195.69
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1,2,3,6-Tetrahydro-4-phenylpyridine Hydrochloride