missing translation for 'onlineSavingsMsg'
Learn More

1,2,3,5-Tetrafluorobenzene 98.0+%, TCI America™

Supplier:  TCI America T18025G

Catalog No. T18025G

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

1,2,3,5-Tetrafluorobenzene
Colorless
C6H2F4
5 g
benzene, 1,2,3,5-tetrafluoro, unii-bv201pz5tv, bv201pz5tv, 1,2,3,5 tetrafluorobenzene, pubchem1062, acmc-1carr, intermediates-zcf02631, 1,2,4,6-tetrafluorobenzene, 1,3,4,5-tetrafluorobenzene, 1,2,3,5,-tetrafluorobenzene
C1=C(C=C(C(=C1F)F)F)F
150.076
150.08
Liquid
2367-82-0
84°C
MFCD00000302
1993
UHHYOKRQTQBKSB-UHFFFAOYSA-N
1,2,3,5-tetrafluorobenzene
16910
≥98.0% (GC)

Chemical Identifiers

2367-82-0
150.076
UHHYOKRQTQBKSB-UHFFFAOYSA-N
16910
C1=C(C=C(C(=C1F)F)F)F
C6H2F4
MFCD00000302
benzene, 1,2,3,5-tetrafluoro, unii-bv201pz5tv, bv201pz5tv, 1,2,3,5 tetrafluorobenzene, pubchem1062, acmc-1carr, intermediates-zcf02631, 1,2,4,6-tetrafluorobenzene, 1,3,4,5-tetrafluorobenzene, 1,2,3,5,-tetrafluorobenzene
1,2,3,5-tetrafluorobenzene
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents

Promotions

Promotions