accessibility menu, dialog, popup

UserName

CAS RN 2367-82-0

FFFF

CAS RN 2367-82-0

IUPAC Name: 1,2,3,5-tetrafluorobenzene
Synonyms: 1,2,3,5-tetrafluorobenzene
Molecular Weight (g/mol): 150.08
Molecular Formula: C6H2F4
InChi Key: UHHYOKRQTQBKSB-UHFFFAOYSA-N
SMILES: FC1=CC(F)=C(F)C(F)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results