missing translation for 'onlineSavingsMsg'
Learn More

1-(2,3,4-Trimethoxybenzyl)piperazine Dihydrochloride 98.0+%, TCI America™

Catalog No. p-7137375
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T27265G 5 g
T272625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T27265G Supplier TCI America Supplier No. T27265G

Chemical Identifiers

CAS 13171-25-0
Molecular Formula C14H24Cl2N2O3
Molecular Weight (g/mol) 339.257
MDL Number MFCD00243086
InChI Key VYFLPFGUVGMBEP-UHFFFAOYSA-N
Synonym trimetazidine dihydrochloride, 1-2,3,4-trimethoxybenzyl piperazine dihydrochloride, kyurinett, trimetazidine hydrochloride, trimetajust, lubomail, vastarel, yosimilon, 1-2,3,4-trimethoxyphenyl methyl piperazine dihydrochloride, trimetazidine 2hcl
PubChem CID 83201
IUPAC Name 1-[(2,3,4-trimethoxyphenyl)methyl]piperazine;dihydrochloride
SMILES COC1=C(C(=C(C=C1)CN2CCNCC2)OC)OC.Cl.Cl

Specifications

Melting Point 228°C
Color White
Quantity 5 g
Formula Weight 339.26
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 1-(2,3,4-Trimethoxybenzyl)piperazine Dihydrochloride
RTECSNumber TM2976100
TSCA No