accessibility menu, dialog, popup

UserName

HHClClH3CONHNOH3COCH3

CAS RN 13171-25-0

IUPAC Name: dihydrogen 1-[(2,3,4-trimethoxyphenyl)methyl]piperazine dichloride
Synonyms: dihydrogen trimetazidine dichloride
Molecular Weight (g/mol): 339.26
Molecular Formula: C14H24Cl2N2O3
InChi Key: VYFLPFGUVGMBEP-UHFFFAOYSA-N
SMILES: [H+].[H+].[Cl-].[Cl-].COC1=CC=C(CN2CCNCC2)C(OC)=C1OC
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results