missing translation for 'onlineSavingsMsg'
Learn More

1,2,3,4-Tetrahydronaphthalene 97.0+%, TCI America™

Supplier:  TCI America T010725ML

 View more versions of this product

Catalog No. T010725ML


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

1,2,3,4-Tetrahydronaphthalene
-36°C
207°C
MFCD00001733
3082
CXWXQJXEFPUFDZ-UHFFFAOYSA-N
1,2,3,4-tetrahydronaphthalene
8404
132.21
Liquid
119-64-2
Yellow
C10H12
25 mL
tetralin, benzocyclohexane, tetrahydronaphthalene, bacticin, tetraline, tetranap, naphthalene, 1,2,3,4-tetrahydro, tetralina, naphthalene 1,2,3,4-tetrahydride, tetralene
C1CCC2=CC=CC=C2C1
132.21
CHEBI:35008
≥97.0% (GC)

Chemical Identifiers

119-64-2
132.21
CXWXQJXEFPUFDZ-UHFFFAOYSA-N
8404
1,2,3,4-tetrahydronaphthalene
C10H12
MFCD00001733
tetralin, benzocyclohexane, tetrahydronaphthalene, bacticin, tetraline, tetranap, naphthalene, 1,2,3,4-tetrahydro, tetralina, naphthalene 1,2,3,4-tetrahydride, tetralene
CHEBI:35008
C1CCC2=CC=CC=C2C1
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (4)-0574

RTECSNumber : QK3850000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions