missing translation for 'onlineSavingsMsg'
Learn More

1,2,3,4-Tetrahydronaphthalene 97.0+%, TCI America™

Catalog No. p-7136085
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T010725ML 25 mL
T0107500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T010725ML Supplier TCI America Supplier No. T010725ML
Only null left

Chemical Identifiers

CAS 119-64-2
Molecular Formula C10H12
Molecular Weight (g/mol) 132.21
MDL Number MFCD00001733
InChI Key CXWXQJXEFPUFDZ-UHFFFAOYSA-N
Synonym tetralin, benzocyclohexane, tetrahydronaphthalene, bacticin, tetraline, tetranap, naphthalene, 1,2,3,4-tetrahydro, tetralina, naphthalene 1,2,3,4-tetrahydride, tetralene
PubChem CID 8404
ChEBI CHEBI:35008
IUPAC Name 1,2,3,4-tetrahydronaphthalene
SMILES C1CCC2=CC=CC=C2C1

Specifications

Melting Point -36°C
Color Yellow
Boiling Point 207°C
Quantity 25 mL
UN Number 3082
Formula Weight 132.21
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,2,3,4-Tetrahydronaphthalene
EINECSNumber (4)-0574
RTECSNumber QK3850000
TSCA Yes