missing translation for 'onlineSavingsMsg'
Learn More

1,2,3,4-Tetrahydronaphthalene-1-carboxylic Acid 98.0+%, TCI America™

Catalog No. p-7137469
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T29035G 5 g
T290325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T29035G Supplier TCI America Supplier No. T29035G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1914-65-4
Molecular Formula C11H12O2
Molecular Weight (g/mol) 176.215
MDL Number MFCD02179127
InChI Key VDLWTJCSPSUGOA-UHFFFAOYSA-N
Synonym 1,2,3,4-tetrahydro-1-naphthoic acid, 1-naphthalenecarboxylic acid, 1,2,3,4-tetrahydro, tetralin-1-carboxylic acid, 1,2,3,4-tetrahedro-naphthoic acid, 1,2,3,4-tetrahydro-naphthalene-1-carboxylic acid, chembl81606, 1-tetralincarboxylic acid, 1,2,3,4-tetrahydro-1-naphthalenecarboxylic acid, 1,2,3,4-tetrahydronaphthalenecarboxylic acid, tetrahydronaphthoic acid
PubChem CID 12598146
IUPAC Name 1,2,3,4-tetrahydronaphthalene-1-carboxylic acid
SMILES C1CC(C2=CC=CC=C2C1)C(=O)O

Specifications

Melting Point 84°C
Boiling Point 135°C
Quantity 5 g
Formula Weight 176.22
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1,2,3,4-Tetrahydronaphthalene-1-carboxylic Acid
EINECSNumber (4)-1181
TSCA No