accessibility menu, dialog, popup

UserName

CAS RN 1914-65-4

IUPAC Name: 1,2,3,4-tetrahydronaphthalene-1-carboxylic acid
Synonyms: 1,2,3,4-tetrahydronaphthalene-1-carboxylic acid
Molecular Weight (g/mol): 176.22
Molecular Formula: C11H12O2
InChi Key: VDLWTJCSPSUGOA-UHFFFAOYNA-N
SMILES: OC(=O)C1CCCC2=CC=CC=C12
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)