missing translation for 'onlineSavingsMsg'
Learn More

1,2,3,4-Tetrahydro-1-naphthoic acid, 98%

Catalog No. p-7031163
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH32138MD 250 mg
AAH3213803 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH32138MD Supplier Thermo Scientific Chemicals Supplier No. H32138MD
Only null left

CAS: 1914-65-4 | C11H12O2 | 176.215 g/mol

1,2,3,4-Tetrahydro-1-naphthoic acid is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. 1,2,3,4-tetrahydronaphthalene-1-carboxylic acid is a reagent used to produce protease inhibitors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,2,3,4-Tetrahydro-1-naphthoic acid is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. 1,2,3,4-tetrahydronaphthalene-1-carboxylic acid is a reagent used to produce protease inhibitors.

Solubility
Soluble in methanol, and chloroform.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.

Chemical Identifiers

CAS 1914-65-4
Molecular Formula C11H12O2
Molecular Weight (g/mol) 176.215
MDL Number MFCD02179127
InChI Key VDLWTJCSPSUGOA-UHFFFAOYSA-N
Synonym 1,2,3,4-tetrahydro-1-naphthoic acid, 1-naphthalenecarboxylic acid, 1,2,3,4-tetrahydro, tetralin-1-carboxylic acid, 1,2,3,4-tetrahedro-naphthoic acid, 1,2,3,4-tetrahydro-naphthalene-1-carboxylic acid, chembl81606, 1-tetralincarboxylic acid, 1,2,3,4-tetrahydro-1-naphthalenecarboxylic acid, 1,2,3,4-tetrahydronaphthalenecarboxylic acid, tetrahydronaphthoic acid
PubChem CID 12598146
IUPAC Name 1,2,3,4-tetrahydronaphthalene-1-carboxylic acid
SMILES C1CC(C2=CC=CC=C2C1)C(=O)O

Specifications

Melting Point 82°C to 85°C
Quantity 250 mg
Solubility Information Soluble in methanol,and chloroform.
Formula Weight 176.22
Percent Purity 98%
Chemical Name or Material 1,2,3,4-Tetrahydro-1-naphthoic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only