missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,2,3,4,5-Pentamethylcyclopentadiene 93.0+%, TCI America™
Supplier: TCI America P129225ML
Specifications
| 1,2,3,4,5-Pentamethylcyclopentadiene | |
| Yellow | |
| C10H16 | |
| 25 mL | |
| 1,2,3,4,5-pentamethylcyclopentadiene, pentamethylcyclopentadiene, cyclopentadiene, 1,2,3,4,5-pentamethyl, 1,3-cyclopentadiene, 1,2,3,4,5-pentamethyl, 1,3-cyclopentadiene, pentamethyl, 1,2,3,4,5-pentamethyl-1,3-cyclopentadiene, pubchem20101, acmc-209je4, wqiqnkqyeumpbm-uhfffaoysa, 1,2,3,4,5-pentamethylcyclopentadien | |
| CC1C(=C(C(=C1C)C)C)C | |
| 136.238 | |
| 136.24 | |
| Liquid |
| 4045-44-7 | |
| 60°C | |
| MFCD00001354 | |
| 3295 | |
| WQIQNKQYEUMPBM-UHFFFAOYSA-N | |
| 1,2,3,4,5-pentamethylcyclopenta-1,3-diene | |
| 77667 | |
| ≥93.0% (GC) |
Chemical Identifiers
| 4045-44-7 | |
| 136.238 | |
| WQIQNKQYEUMPBM-UHFFFAOYSA-N | |
| 77667 | |
| CC1C(=C(C(=C1C)C)C)C |
| C10H16 | |
| MFCD00001354 | |
| 1,2,3,4,5-pentamethylcyclopentadiene, pentamethylcyclopentadiene, cyclopentadiene, 1,2,3,4,5-pentamethyl, 1,3-cyclopentadiene, 1,2,3,4,5-pentamethyl, 1,3-cyclopentadiene, pentamethyl, 1,2,3,4,5-pentamethyl-1,3-cyclopentadiene, pubchem20101, acmc-209je4, wqiqnkqyeumpbm-uhfffaoysa, 1,2,3,4,5-pentamethylcyclopentadien | |
| 1,2,3,4,5-pentamethylcyclopenta-1,3-diene |
Safety and Handling
TSCA : No