accessibility menu, dialog, popup

UserName

CH3CH3H3CH3CCH3

CAS RN 4045-44-7

IUPAC Name: 1,2,3,4,5-pentamethylcyclopenta-1,3-diene
Synonyms: pentamethylcyclopentadiene
Molecular Weight (g/mol): 136.24
Molecular Formula: C10H16
InChi Key: WQIQNKQYEUMPBM-UHFFFAOYSA-N
SMILES: CC1C(C)=C(C)C(C)=C1C
Boiling Point 
  • (2)
  • (1)
  • (2)
Molecular Weight (g/mol) 
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (3)
  • (2)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (4)