missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,2,3,4,5-Pentamethylcyclopentadiene 93.0+%, TCI America™
$253.51 - $900.33
Chemical Identifiers
| CAS | 4045-44-7 |
|---|---|
| Molecular Formula | C10H16 |
| Molecular Weight (g/mol) | 136.238 |
| MDL Number | MFCD00001354 |
| InChI Key | WQIQNKQYEUMPBM-UHFFFAOYSA-N |
| Synonym | 1,2,3,4,5-pentamethylcyclopentadiene, pentamethylcyclopentadiene, cyclopentadiene, 1,2,3,4,5-pentamethyl, 1,3-cyclopentadiene, 1,2,3,4,5-pentamethyl, 1,3-cyclopentadiene, pentamethyl, 1,2,3,4,5-pentamethyl-1,3-cyclopentadiene, pubchem20101, acmc-209je4, wqiqnkqyeumpbm-uhfffaoysa, 1,2,3,4,5-pentamethylcyclopentadien |
| PubChem CID | 77667 |
| IUPAC Name | 1,2,3,4,5-pentamethylcyclopenta-1,3-diene |
| SMILES | CC1C(=C(C(=C1C)C)C)C |
Chemical Identifiers
| 4045-44-7 | |
| 136.238 | |
| WQIQNKQYEUMPBM-UHFFFAOYSA-N | |
| 77667 | |
| CC1C(=C(C(=C1C)C)C)C |
| C10H16 | |
| MFCD00001354 | |
| 1,2,3,4,5-pentamethylcyclopentadiene, pentamethylcyclopentadiene, cyclopentadiene, 1,2,3,4,5-pentamethyl, 1,3-cyclopentadiene, 1,2,3,4,5-pentamethyl, 1,3-cyclopentadiene, pentamethyl, 1,2,3,4,5-pentamethyl-1,3-cyclopentadiene, pubchem20101, acmc-209je4, wqiqnkqyeumpbm-uhfffaoysa, 1,2,3,4,5-pentamethylcyclopentadien | |
| 1,2,3,4,5-pentamethylcyclopenta-1,3-diene |
Specifications
| 4045-44-7 | |
| 60°C | |
| MFCD00001354 | |
| 3295 | |
| WQIQNKQYEUMPBM-UHFFFAOYSA-N | |
| 1,2,3,4,5-pentamethylcyclopenta-1,3-diene | |
| 77667 | |
| ≥93.0% (GC) | |
| 1,2,3,4,5-Pentamethylcyclopentadiene |
| Yellow | |
| C10H16 | |
| 5 mL | |
| 1,2,3,4,5-pentamethylcyclopentadiene, pentamethylcyclopentadiene, cyclopentadiene, 1,2,3,4,5-pentamethyl, 1,3-cyclopentadiene, 1,2,3,4,5-pentamethyl, 1,3-cyclopentadiene, pentamethyl, 1,2,3,4,5-pentamethyl-1,3-cyclopentadiene, pubchem20101, acmc-209je4, wqiqnkqyeumpbm-uhfffaoysa, 1,2,3,4,5-pentamethylcyclopentadien | |
| CC1C(=C(C(=C1C)C)C)C | |
| 136.238 | |
| 136.24 | |
| Liquid |
Safety and Handling
TSCA : No