missing translation for 'onlineSavingsMsg'
Learn More

1,2,3,4,5,6,7,8-Octahydrophenanthrene 98.0+%, TCI America™

Catalog No. O00215G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O00215G 5 g
O00211G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. O00215G Supplier TCI America Supplier No. O00215G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 5325-97-3
Molecular Formula C14H18
Molecular Weight (g/mol) 186.30
MDL Number MFCD00058943
InChI Key YSZLFGZFQTTZIQ-UHFFFAOYSA-N
PubChem CID 79221
IUPAC Name 1,2,3,4,5,6,7,8-octahydrophenanthrene
SMILES C1CCC2=C(C1)C=CC1=C2CCCC1

Specifications

Color Yellow
Boiling Point 295°C
Quantity 5 g
Formula Weight 186.30
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,2,3,4,5,6,7,8-Octahydrophenanthrene