accessibility menu, dialog, popup

UserName

CAS RN 5325-97-3

IUPAC Name: 1,2,3,4,5,6,7,8-octahydrophenanthrene
Synonyms: 1,2,3,4,5,6,7,8-octahydrophenanthrene
Molecular Weight (g/mol): 186.3
Molecular Formula: C14H18
InChi Key: YSZLFGZFQTTZIQ-UHFFFAOYSA-N
SMILES: C1CCC2=C(C1)C=CC1=C2CCCC1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results