missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,2,3,4,5,6,7,8-Octahydrophenanthrene 98.0+%, TCI America™
Specifications
Specifications
| Color | Yellow |
| Boiling Point | 295°C |
| Quantity | 1 g |
| Formula Weight | 186.30 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | 1,2,3,4,5,6,7,8-Octahydrophenanthrene |
Safety and Handling
| TSCA | No |