missing translation for 'onlineSavingsMsg'
Learn More

1,18-Octadecanedicarboxylic acid

CAS: 2424-92-2 | C20H38O4 | 342.52 g/mol

$81.12 - $3,523.70

Chemical Identifiers

CAS 2424-92-2
Molecular Formula C20H38O4
Molecular Weight (g/mol) 342.52
MDL Number MFCD00059627
InChI Key JJOJFIHJIRWASH-UHFFFAOYSA-N
Synonym eicosanedioic acid, unii-sod235tgnz, 1,18-octadecanedicarboxylic acid, sod235tgnz, octadecane-1,18-dicarboxylic acid, eicosanedioicacid, 1,20-icosanedioic acid, 1,20-eicosanedioic acid, eicosa-1,20-dioic acid
PubChem CID 75502
ChEBI CHEBI:73728
IUPAC Name icosanedioic acid
SMILES C(CCCCCCCCCC(=O)O)CCCCCCCCC(=O)O
View More Specs

Products 4
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA4200404
View Documents
Thermo Scientific Chemicals
04200404
2 g N/A
Only null left
 
AA4200409
View Documents
Thermo Scientific Chemicals
04200409
10 g N/A
Only null left
 
AA4200418
View Documents
Thermo Scientific Chemicals
04200418
50 g N/A
Only null left
 
AA4200430
View Documents
Thermo Scientific Chemicals
04200430
250 g N/A
Only null left
 
Description

Description

1,18-Octadecanedicarboxylic acid is used as an intermediate in organic synthesis, as dispersing agent, emulsifying agent, as lubricant.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

2424-92-2
342.52
JJOJFIHJIRWASH-UHFFFAOYSA-N
75502
icosanedioic acid
C20H38O4
MFCD00059627
eicosanedioic acid, unii-sod235tgnz, 1,18-octadecanedicarboxylic acid, sod235tgnz, octadecane-1,18-dicarboxylic acid, eicosanedioicacid, 1,20-icosanedioic acid, 1,20-eicosanedioic acid, eicosa-1,20-dioic acid
CHEBI:73728
C(CCCCCCCCCC(=O)O)CCCCCCCCC(=O)O

Specifications

2424-92-2
C20H38O4
MFCD00059627
eicosanedioic acid, unii-sod235tgnz, 1,18-octadecanedicarboxylic acid, sod235tgnz, octadecane-1,18-dicarboxylic acid, eicosanedioicacid, 1,20-icosanedioic acid, 1,20-eicosanedioic acid, eicosa-1,20-dioic acid
JJOJFIHJIRWASH-UHFFFAOYSA-N
icosanedioic acid
75502
342.52
1,18-Octadecanedicarboxylic acid
127°C
HO2C(CH2)18CO2H
2 g
Slightly soluble in water.
C(CCCCCCCCCC(=O)O)CCCCCCCCC(=O)O
342.52
CHEBI:73728
Powder
Videos
Safety and Handling

Safety and Handling

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only