missing translation for 'onlineSavingsMsg'
Learn More

1,18-Octadecanedicarboxylic acid

CAS: 2424-92-2 | C20H38O4 | 342.52 g/mol

Supplier:  Thermo Scientific Chemicals 04200418

 View more versions of this product

Catalog No. AA4200418


Only null left

Description

Description

1,18-Octadecanedicarboxylic acid is used as an intermediate in organic synthesis, as dispersing agent, emulsifying agent, as lubricant.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

1,18-Octadecanedicarboxylic acid
127°C
HO2C(CH2)18CO2H
50 g
Slightly soluble in water.
C(CCCCCCCCCC(=O)O)CCCCCCCCC(=O)O
342.52
CHEBI:73728
Powder
2424-92-2
C20H38O4
MFCD00059627
eicosanedioic acid, unii-sod235tgnz, 1,18-octadecanedicarboxylic acid, sod235tgnz, octadecane-1,18-dicarboxylic acid, eicosanedioicacid, 1,20-icosanedioic acid, 1,20-eicosanedioic acid, eicosa-1,20-dioic acid
JJOJFIHJIRWASH-UHFFFAOYSA-N
icosanedioic acid
75502
342.52

Chemical Identifiers

2424-92-2
342.52
JJOJFIHJIRWASH-UHFFFAOYSA-N
75502
icosanedioic acid
C20H38O4
MFCD00059627
eicosanedioic acid, unii-sod235tgnz, 1,18-octadecanedicarboxylic acid, sod235tgnz, octadecane-1,18-dicarboxylic acid, eicosanedioicacid, 1,20-icosanedioic acid, 1,20-eicosanedioic acid, eicosa-1,20-dioic acid
CHEBI:73728
C(CCCCCCCCCC(=O)O)CCCCCCCCC(=O)O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only