missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,12-Dibromododecane 98.0+%, TCI America™
$121.28 - $1172.43
Chemical Identifiers
| CAS | 3344-70-5 |
|---|---|
| Molecular Formula | C12H24Br2 |
| Molecular Weight (g/mol) | 328.132 |
| MDL Number | MFCD00000226 |
| InChI Key | ZJJATABWMGVVRZ-UHFFFAOYSA-N |
| Synonym | dodecamethylene dibromide, dodecane, 1,12-dibromo, alpha,omega-dibromododecane, labotest-bb ltbb002671, pubchem3879, 1, 12-dibromododecane, 1,12-dibromo-dodecane, 1,12-bis bromanyl dodecane, 4-01-00-00503 beilstein handbook reference, ksc225m2h |
| PubChem CID | 18766 |
| IUPAC Name | 1,12-dibromododecane |
| SMILES | C(CCCCCCBr)CCCCCBr |
Chemical Identifiers
| 3344-70-5 | |
| 328.132 | |
| ZJJATABWMGVVRZ-UHFFFAOYSA-N | |
| 18766 | |
| C(CCCCCCBr)CCCCCBr |
| C12H24Br2 | |
| MFCD00000226 | |
| dodecamethylene dibromide, dodecane, 1,12-dibromo, alpha,omega-dibromododecane, labotest-bb ltbb002671, pubchem3879, 1, 12-dibromododecane, 1,12-dibromo-dodecane, 1,12-bis bromanyl dodecane, 4-01-00-00503 beilstein handbook reference, ksc225m2h | |
| 1,12-dibromododecane |
Specifications
| 3344-70-5 | |
| White-Yellow | |
| C12H24Br2 | |
| 25 g | |
| ZJJATABWMGVVRZ-UHFFFAOYSA-N | |
| 1,12-dibromododecane | |
| 18766 | |
| ≥98.0% (GC) | |
| 1,12-Dibromododecane |
| 40°C | |
| 215°C | |
| MFCD00000226 | |
| dodecamethylene dibromide, dodecane, 1,12-dibromo, alpha,omega-dibromododecane, labotest-bb ltbb002671, pubchem3879, 1, 12-dibromododecane, 1,12-dibromo-dodecane, 1,12-bis bromanyl dodecane, 4-01-00-00503 beilstein handbook reference, ksc225m2h | |
| C(CCCCCCBr)CCCCCBr | |
| 328.132 | |
| 328.13 | |
| Crystalline Powder |
Safety and Handling
RTECSNumber : JR2295000
TSCA : Yes
Recommended Storage : Refrigerator