accessibility menu, dialog, popup

UserName

CAS RN 3344-70-5

IUPAC Name: 1,12-dibromododecane
Synonyms: dodecane, 1,12-dibromo-
Molecular Weight (g/mol): 328.13
Molecular Formula: C12H24Br2
InChi Key: ZJJATABWMGVVRZ-UHFFFAOYSA-N
SMILES: BrCCCCCCCCCCCCBr
Boiling Point 
  • (3)
  • (3)
  • (3)
Molecular Weight (g/mol) 
  • (3)
  • (6)
Percent Purity 
  • (3)
  • (3)
  • (3)
Quantity 
  • (3)
  • (3)
  • (1)
  • (2)
Physical Form 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (3)
  • (6)
  • (3)
  • (3)
  • (3)
  • (3)
  • (6)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (2)
  • (3)
  • (3)