missing translation for 'onlineSavingsMsg'
Learn More

1,12-Dibromododecane, 96%

Catalog No. AC158450250
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC158450250 25 g Glass bottle
AC158451000 100 g Glass bottle
AC158455000 500 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC158450250 Supplier Thermo Scientific Chemicals Supplier No. 158450250

CAS: 3344-70-5 | C12H24Br2 | 328.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3344-70-5
Molecular Formula C12H24Br2
Molecular Weight (g/mol) 328.13
MDL Number MFCD00000226
InChI Key ZJJATABWMGVVRZ-UHFFFAOYSA-N
Synonym dodecamethylene dibromide, dodecane, 1,12-dibromo, alpha,omega-dibromododecane, labotest-bb ltbb002671, pubchem3879, 1, 12-dibromododecane, 1,12-dibromo-dodecane, 1,12-bis bromanyl dodecane, 4-01-00-00503 beilstein handbook reference, ksc225m2h
PubChem CID 18766
IUPAC Name 1,12-dibromododecane
SMILES C(CCCCCCBr)CCCCCBr

Specifications

CAS Min % 95.0
CAS Max % 100.0
Melting Point 37.0°C to 42.0°C
Color White to Brown
Boiling Point 215.0°C (15.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 96%
Packaging Glass bottle
Linear Formula Br(CH2)12Br
Quantity 25 g
Beilstein 01, II, 134
Formula Weight 328.13
Percent Purity 96%
Physical Form Crystalline Solid
Chemical Name or Material 1,12-Dibromododecane
Show More Show Less
EINECSNumber 222-096-5

RUO – Research Use Only