missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,11-Dibromoundecane 96.0+%, TCI America™
$46.20 - $127.07
Chemical Identifiers
| CAS | 16696-65-4 |
|---|---|
| Molecular Formula | C11H22Br2 |
| Molecular Weight (g/mol) | 314.105 |
| MDL Number | MFCD00000224 |
| InChI Key | SIBVHGAPHVRHMJ-UHFFFAOYSA-N |
| Synonym | undecamethylene dibromide, undecane, 1,11-dibromo, 1,11-dibromo-undecane, acmc-1buwu, ksc177k0h, 1,11-dibromo undecane, 1,11-dibromoundecane, 1,11-dibromoundecane, purum gc |
| PubChem CID | 85551 |
| IUPAC Name | 1,11-dibromoundecane |
| SMILES | C(CCCCCBr)CCCCCBr |
Chemical Identifiers
| 16696-65-4 | |
| 314.105 | |
| SIBVHGAPHVRHMJ-UHFFFAOYSA-N | |
| 85551 | |
| C(CCCCCBr)CCCCCBr |
| C11H22Br2 | |
| MFCD00000224 | |
| undecamethylene dibromide, undecane, 1,11-dibromo, 1,11-dibromo-undecane, acmc-1buwu, ksc177k0h, 1,11-dibromo undecane, 1,11-dibromoundecane, 1,11-dibromoundecane, purum gc | |
| 1,11-dibromoundecane |
Specifications
| 16696-65-4 | |
| Yellow | |
| C11H22Br2 | |
| 1 g | |
| SIBVHGAPHVRHMJ-UHFFFAOYSA-N | |
| 1,11-dibromoundecane | |
| 85551 | |
| ≥96.0% (GC) | |
| 1,11-Dibromoundecane |
| -11°C | |
| 192°C | |
| MFCD00000224 | |
| undecamethylene dibromide, undecane, 1,11-dibromo, 1,11-dibromo-undecane, acmc-1buwu, ksc177k0h, 1,11-dibromo undecane, 1,11-dibromoundecane, 1,11-dibromoundecane, purum gc | |
| C(CCCCCBr)CCCCCBr | |
| 314.105 | |
| 314.11 | |
| Liquid |
Safety and Handling
TSCA : No