CAS RN 16696-65-4
IUPAC Name:
1,11-dibromoundecane
Synonyms:
1,11-dibromo-undecane
Molecular Weight (g/mol):
314.11
Molecular Formula:
C11H22Br2
InChi Key:
SIBVHGAPHVRHMJ-UHFFFAOYSA-N
SMILES:
BrCCCCCCCCCCCBr
1
–
1
of
1
results
1
1,11-Dibromoundecane 96.0+%, TCI America™
CAS: 16696-65-4 Molecular Formula: C11H22Br2 Molecular Weight (g/mol): 314.105 MDL Number: MFCD00000224 InChI Key: SIBVHGAPHVRHMJ-UHFFFAOYSA-N Synonym: undecamethylene dibromide,undecane, 1,11-dibromo,1,11-dibromo-undecane,acmc-1buwu,ksc177k0h,1,11-dibromo undecane,1,11-dibromoundecane,1,11-dibromoundecane, purum gc PubChem CID: 85551 IUPAC Name: 1,11-dibromoundecane SMILES: C(CCCCCBr)CCCCCBr