missing translation for 'onlineSavingsMsg'
Learn More

1,11-Dibromoundecane 96.0+%, TCI America™

Catalog No. D24875G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D24875G 5 g
D24871G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D24875G Supplier TCI America Supplier No. D24875G
Only null left

Chemical Identifiers

CAS 16696-65-4
Molecular Formula C11H22Br2
Molecular Weight (g/mol) 314.105
MDL Number MFCD00000224
InChI Key SIBVHGAPHVRHMJ-UHFFFAOYSA-N
Synonym undecamethylene dibromide, undecane, 1,11-dibromo, 1,11-dibromo-undecane, acmc-1buwu, ksc177k0h, 1,11-dibromo undecane, 1,11-dibromoundecane, 1,11-dibromoundecane, purum gc
PubChem CID 85551
IUPAC Name 1,11-dibromoundecane
SMILES C(CCCCCBr)CCCCCBr

Specifications

Melting Point -11°C
Color Yellow
Boiling Point 192°C
Quantity 5 g
Formula Weight 314.11
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,11-Dibromoundecane