missing translation for 'onlineSavingsMsg'
Learn More

1,10-Phenanthroline, 0.3% w/v Acidified, Reagents

Catalog No. 01334284
Change view
Click to view available options
Quantity:
500 mL
1 L
20 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01334284 20 L
01334285 1 L
01334283 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01334284 Supplier Labchem Inc Supplier No. CP12180020F

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

1,10-Phenanthroline, 0.3% w/v Acidified, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7732-18-5 , 5144-89-8 , 7647-01-0
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym o-Phenanthroline
IUPAC Name water
SMILES O

Specifications

Color Colorless
pH 1.9
Boiling Point 100°C
Packaging Cubitainer
Quantity 20 L
Specific Gravity 1.001
Percent Purity 0.3%
Grade Reagent
Physical Form Liquid
Chemical Name or Material 1,10-Phenanthroline, 0.3% w/v Acidified