missing translation for 'onlineSavingsMsg'
Learn More

1,10-Dibromodecane, 97%

Catalog No. AC112780250
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC112780250 25 mL Glass bottle
AC112781000 100 mL Glass bottle
AC112785000 500 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC112780250 Supplier Thermo Scientific Chemicals Supplier No. 112780250

CAS: 4101-68-2 | C10H20Br2 | 300.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4101-68-2
Molecular Formula C10H20Br2
Molecular Weight (g/mol) 300.08
MDL Number MFCD00000222
InChI Key GTQHJCOHNAFHRE-UHFFFAOYSA-N
Synonym decamethylene dibromide, decane, 1,10-dibromo, 1,10-dibromodecan, 1,10-dibromo-decan, 1,10-decamethylene bromide, 1,10-decamethylene dibromide, a,w-dibromodecane, 1,10-dibromdecane, 1,10 dibromodecane, decane,10-dibromo
PubChem CID 221483
IUPAC Name 1,10-dibromodecane
SMILES BrCCCCCCCCCCBr

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 25°C to 30°C
Color Beige to Brown
Density 1.3400g/mL
Boiling Point 160°C (15.0 mmHg)
Flash Point >112°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.4912
Linear Formula Br(CH2)10Br
Quantity 25 mL
Beilstein 01, I, 64
Specific Gravity 1.34
Solubility Information Solubility in water: insoluble
Formula Weight 300.07
Percent Purity 97%
Physical Form Crystalline Low Melting Solid
Chemical Name or Material 1, 10-Dibromodecane
Show More Show Less
EINECSNumber 223-871-
Recommended Storage Product may darken during storage

RUO – Research Use Only