accessibility menu, dialog, popup

UserName

BrBr

CAS RN 4101-68-2

IUPAC Name: 1,10-dibromodecane
Synonyms: 1,10-dibromodecane
Molecular Weight (g/mol): 300.08
Molecular Formula: C10H20Br2
InChi Key: GTQHJCOHNAFHRE-UHFFFAOYSA-N
SMILES: BrCCCCCCCCCCBr
Percent Purity 
  • (2)
  • (5)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)
  • (3)
Physical Form 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (5)
  • (2)
  • (5)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (3)
  • (2)