missing translation for 'onlineSavingsMsg'
Learn More

1,10-Dibromodecane, 97%

CAS: 4101-68-2 | C10H20Br2 | 300.08 g/mol

$211.84 - $699.05

Chemical Identifiers

CAS 4101-68-2
Molecular Formula C10H20Br2
Molecular Weight (g/mol) 300.08
MDL Number MFCD00000222
InChI Key GTQHJCOHNAFHRE-UHFFFAOYSA-N
Synonym decamethylene dibromide, decane, 1,10-dibromo, 1,10-dibromodecan, 1,10-dibromo-decan, 1,10-decamethylene bromide, 1,10-decamethylene dibromide, a,w-dibromodecane, 1,10-dibromdecane, 1,10 dibromodecane, decane,10-dibromo
PubChem CID 221483
IUPAC Name 1,10-dibromodecane
SMILES BrCCCCCCCCCCBr
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1235722
View Documents
Thermo Scientific Chemicals
A1235722
100 g
Each for $211.84
Only null left
 
AAA1235736
View Documents
Thermo Scientific Chemicals
A1235736
500 g
Each for $699.05
Only null left
 
Description

Description

1,10-Dibromodecane is used as an alkylating agent and as synthetic organic intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,10-Dibromodecane is used as an alkylating agent and as synthetic organic intermediate.

Solubility
Insoluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sea
Specifications

Chemical Identifiers

4101-68-2
300.08
GTQHJCOHNAFHRE-UHFFFAOYSA-N
221483
BrCCCCCCCCCCBr
C10H20Br2
MFCD00000222
decamethylene dibromide, decane, 1,10-dibromo, 1,10-dibromodecan, 1,10-dibromo-decan, 1,10-decamethylene bromide, 1,10-decamethylene dibromide, a,w-dibromodecane, 1,10-dibromdecane, 1,10 dibromodecane, decane,10-dibromo
1,10-dibromodecane

Specifications

4101-68-2
1.34
>110°C (230°F)
1.493
100 g
decamethylene dibromide, decane, 1,10-dibromo, 1,10-dibromodecan, 1,10-dibromo-decan, 1,10-decamethylene bromide, 1,10-decamethylene dibromide, a,w-dibromodecane, 1,10-dibromdecane, 1,10 dibromodecane, decane,10-dibromo
GTQHJCOHNAFHRE-UHFFFAOYSA-N
1,10-dibromodecane
221483
97%
26°C to 30°C
157°C to 159°C (14 mmHg)
C10H20Br2
MFCD00000222
506156
Insoluble in water.
BrCCCCCCCCCCBr
300.08
300.08
1,10-Dibromodecane
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335;
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a;
Avoid breathing dust/fume/gas/mist/vapours/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

EINECSNumber : 223-871-0

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only