missing translation for 'onlineSavingsMsg'
Learn More

1,10-Bis(4-carboxyphenoxy)decane 97.0+%, TCI America™

Catalog No. B14995G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B14995G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B14995G Supplier TCI America Supplier No. B14995G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 74774-61-1
Molecular Formula C24H30O6
Molecular Weight (g/mol) 414.498
MDL Number MFCD00142356
InChI Key XRDKWFXOXXUQJS-UHFFFAOYSA-N
Synonym p,p′C-Dicarboxy-1,10-diphenoxydecane
PubChem CID 400760
IUPAC Name 4-[10-(4-carboxyphenoxy)decoxy]benzoic acid
SMILES C1=CC(=CC=C1C(=O)O)OCCCCCCCCCCOC2=CC=C(C=C2)C(=O)O

Specifications

Melting Point 274°C
Color White-Yellow
Quantity 5 g
Formula Weight 414.50
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 1,10-Bis(4-carboxyphenoxy)decane