accessibility menu, dialog, popup

UserName

OHOOOOOH

CAS RN 74774-61-1

IUPAC Name: 4-{[10-(4-carboxyphenoxy)decyl]oxy}benzoic acid
Synonyms: 4-{[10-(4-carboxyphenoxy)decyl]oxy}benzoic acid
Molecular Weight (g/mol): 414.5
Molecular Formula: C24H30O6
InChi Key: XRDKWFXOXXUQJS-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=C(OCCCCCCCCCCOC2=CC=C(C=C2)C(O)=O)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results