missing translation for 'onlineSavingsMsg'
Learn More

1-(1-Naphthyl)-2-thiourea 98.0+%, TCI America™

Catalog No. N00715G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N00715G 5 g
N007125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N00715G Supplier TCI America Supplier No. N00715G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 86-88-4
Molecular Formula C11H10N2S
Molecular Weight (g/mol) 202.275
MDL Number MFCD00041824
InChI Key PIVQQUNOTICCSA-UHFFFAOYSA-N
Synonym 1-1-naphthyl-2-thiourea, antu, 1-naphthylthiourea, dirax, anturat, rattrack, smeesana, alrato, bantu, kripid
PubChem CID 736366
ChEBI CHEBI:82256
IUPAC Name naphthalen-1-ylthiourea
SMILES C1=CC=C2C(=C1)C=CC=C2NC(=S)N

Specifications

Melting Point 191°C
Color White-Yellow
Quantity 5 g
UN Number 1651
Formula Weight 202.28
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 1-(1-Naphthyl)-2-thiourea
RTECSNumber YT9275000
TSCA Yes