CAS RN 86-88-4
IUPAC Name:
(naphthalen-1-yl)thiourea
Synonyms:
α-naphthylthiourea antu (iso) N-1-napthylthiourea
Molecular Weight (g/mol):
202.28
Molecular Formula:
C11H10N2S
InChi Key:
PIVQQUNOTICCSA-UHFFFAOYSA-N
SMILES:
NC(=S)NC1=C2C=CC=CC2=CC=C1
1
–
1
of
1
results
1
1-(1-Naphthyl)-2-thiourea 98.0+%, TCI America™
CAS: 86-88-4 Molecular Formula: C11H10N2S Molecular Weight (g/mol): 202.275 MDL Number: MFCD00041824 InChI Key: PIVQQUNOTICCSA-UHFFFAOYSA-N Synonym: 1-1-naphthyl-2-thiourea,antu,1-naphthylthiourea,dirax,anturat,rattrack,smeesana,alrato,bantu,kripid PubChem CID: 736366 ChEBI: CHEBI:82256 IUPAC Name: naphthalen-1-ylthiourea SMILES: C1=CC=C2C(=C1)C=CC=C2NC(=S)N