missing translation for 'onlineSavingsMsg'
Learn More

1,1'-Methylenedi-2-naphthol 97.0+%, TCI America™

Catalog No. M141025G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M141025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M141025G Supplier TCI America Supplier No. M141025G
Only null left

Chemical Identifiers

CAS 1096-84-0
Molecular Formula C21H16O2
Molecular Weight (g/mol) 300.357
MDL Number MFCD00046470
InChI Key ZPANWZBSGMDWON-UHFFFAOYSA-N
Synonym 1,1'-methylenedi-2-naphthol, squoxin, 1,1'-methylenebis naphthalen-2-ol, 2-naphthalenol, 1,1'-methylenebis, 1,1'-methylenebis 2-naphthol, 1,1-methylenedi-2-naphthol, unii-wse97k4l90, 2-naphthol, 1,1'-methylenedi, bis 2-hydroxy-1-naphthyl methane, 1-2-hydroxy-1-naphthyl methyl naphthalen-2-ol
PubChem CID 14187
IUPAC Name 1-[(2-hydroxynaphthalen-1-yl)methyl]naphthalen-2-ol
SMILES C1=CC=C2C(=C1)C=CC(=C2CC3=C(C=CC4=CC=CC=C43)O)O

Specifications

Quantity 25 g
Formula Weight 300.36
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,1′-Methylenedi-2-naphthol
EINECSNumber (4)-1616
RTECSNumber QL4400000
TSCA Yes