accessibility menu, dialog, popup

UserName

OHHO

CAS RN 1096-84-0

IUPAC Name: 1-[(2-hydroxynaphthalen-1-yl)methyl]naphthalen-2-ol
Synonyms: squoxin
Molecular Weight (g/mol): 300.36
Molecular Formula: C21H16O2
InChi Key: ZPANWZBSGMDWON-UHFFFAOYSA-N
SMILES: OC1=CC=C2C=CC=CC2=C1CC1=C2C=CC=CC2=CC=C1O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results