missing translation for 'onlineSavingsMsg'
Learn More

1,1-Diphenylethylene oxide

Catalog No. p-3598229
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC391030010 1 g Glass bottle
AC391030050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC391030010 Supplier Thermo Scientific Chemicals Supplier No. 391030010

CAS: 882-59-7 | C14H12O | 196.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 882-59-7
Molecular Formula C14H12O
Molecular Weight (g/mol) 196.25
MDL Number MFCD00040725
InChI Key PRLJMHVNHLTQJJ-UHFFFAOYSA-N
Synonym 1,1-diphenylethylene oxide, oxirane, 2,2-diphenyl, diphenyloxirane, 2,2-diphenyl-oxirane, 2,2-diphenyl oxirane, #, 1,1-diphenyl-ethylenoxide, 1,1-diphenyl-ethylene oxide, 1,1-diphenyl-1,2-epoxyethane
PubChem CID 93564
IUPAC Name 2,2-diphenyloxirane
SMILES C1C(O1)(C2=CC=CC=C2)C3=CC=CC=C3

Specifications

Melting Point 54°C to 56°C
Color White
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 1 g
Formula Weight 196.25
Percent Purity 96%
Physical Form Crystals
Chemical Name or Material 1, 1-Diphenylethylene oxide
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Flammable solid.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep container tightly closed.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 212-929-

RUO – Research Use Only