missing translation for 'onlineSavingsMsg'
Learn More

1,1-Diphenyl-2-thiourea 98.0+%, TCI America™

Catalog No. p-7126501
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D09185G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D09185G Supplier TCI America Supplier No. D09185G
Only null left

Chemical Identifiers

CAS 3898-08-6
Molecular Formula C13H12N2S
Molecular Weight (g/mol) 228.31
MDL Number MFCD00059151
InChI Key FPZXQVCYHDMIIA-UHFFFAOYSA-N
Synonym N,N-Diphenylthiourea
PubChem CID 3034143
IUPAC Name 1,1-diphenylthiourea
SMILES C1=CC=C(C=C1)N(C2=CC=CC=C2)C(=S)N

Specifications

Color White-Yellow
Quantity 5 g
Formula Weight 228.31
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1,1-Diphenyl-2-thiourea
RTECSNumber YT2100000
TSCA Yes