accessibility menu, dialog, popup

UserName

CAS RN 3898-08-6

IUPAC Name: 1,1-diphenylthiourea
Synonyms: thiourea, N,N-diphenyl-
Molecular Weight (g/mol): 228.31
Molecular Formula: C13H12N2S
InChi Key: FPZXQVCYHDMIIA-UHFFFAOYSA-N
SMILES: NC(=S)N(C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results