missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,1-Dimethylguanidine Sulfate 98.0+%, TCI America™
Supplier: TCI America D17995G
Specifications
| 1,1-Dimethylguanidine Sulfate | |
| White | |
| MFCD00013131 | |
| 1,1-dimethylguanidine sulfate 2:1, guanidine, n,n-dimethyl-, sulfate 2:1, bis dimethylguanidine ; sulfuric acid, 1,1-dimethylguanidine hemisulfate, pubchem19198, guanidine,n,n-dimethyl, 1,1-dimethyl guanidinium sulfate, 1,1-dimethylguanidine sulfate salt, 1,1-dimethylguanidinium sulphate 2:1 | |
| CN(C)C(=N)N.CN(C)C(=N)N.OS(=O)(=O)O | |
| 272.324 | |
| 272.32 | |
| Crystalline Powder |
| 598-65-2 | |
| C6H20N6O4S | |
| 5 g | |
| QSCHFHVDZCPIKX-UHFFFAOYSA-N | |
| 1,1-dimethylguanidine;sulfuric acid | |
| 69024 | |
| ≥98.0% (T) |
Chemical Identifiers
| 598-65-2 | |
| 272.324 | |
| QSCHFHVDZCPIKX-UHFFFAOYSA-N | |
| 69024 | |
| CN(C)C(=N)N.CN(C)C(=N)N.OS(=O)(=O)O |
| C6H20N6O4S | |
| MFCD00013131 | |
| 1,1-dimethylguanidine sulfate 2:1, guanidine, n,n-dimethyl-, sulfate 2:1, bis dimethylguanidine ; sulfuric acid, 1,1-dimethylguanidine hemisulfate, pubchem19198, guanidine,n,n-dimethyl, 1,1-dimethyl guanidinium sulfate, 1,1-dimethylguanidine sulfate salt, 1,1-dimethylguanidinium sulphate 2:1 | |
| 1,1-dimethylguanidine;sulfuric acid |
Safety and Handling
TSCA : Yes