accessibility menu, dialog, popup

UserName

CAS RN 598-65-2

IUPAC Name: N,N-dimethylguanidine; sulfuric acid
Synonyms: dimethylguanidine; sulfuric acid
Molecular Weight (g/mol): 185.2
Molecular Formula: C3H11N3O4S
InChi Key: ACPFLXJEPKOPGF-UHFFFAOYSA-N
SMILES: OS(O)(=O)=O.CN(C)C(N)=N
Molecular Weight (g/mol) 
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (3)
Physical Form 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)