missing translation for 'onlineSavingsMsg'
Learn More

1,1-Dimethylguanidine Sulfate 98.0+%, TCI America™

Catalog No. p-7126934
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D17995G 5 g
D179925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D17995G Supplier TCI America Supplier No. D17995G
Only null left

Chemical Identifiers

CAS 598-65-2
Molecular Formula C6H20N6O4S
Molecular Weight (g/mol) 272.324
MDL Number MFCD00013131
InChI Key QSCHFHVDZCPIKX-UHFFFAOYSA-N
Synonym 1,1-dimethylguanidine sulfate 2:1, guanidine, n,n-dimethyl-, sulfate 2:1, bis dimethylguanidine ; sulfuric acid, 1,1-dimethylguanidine hemisulfate, pubchem19198, guanidine,n,n-dimethyl, 1,1-dimethyl guanidinium sulfate, 1,1-dimethylguanidine sulfate salt, 1,1-dimethylguanidinium sulphate 2:1
PubChem CID 69024
IUPAC Name 1,1-dimethylguanidine;sulfuric acid
SMILES CN(C)C(=N)N.CN(C)C(=N)N.OS(=O)(=O)O

Specifications

Color White
Quantity 5 g
Formula Weight 272.32
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 1,1-Dimethylguanidine Sulfate